Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 8-Anilino-1-naphthalenesulfonic acid ammonium salt hydrate

Catalog No. 2169095G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-690-95G 5 g
21-690-9250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 21-690-95G Supplier Sigma Aldrich Supplier No. 2169095G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

8-Anilino-1-naphthalenesulfonic acid ammonium salt hydrate

Specifications

CAS 206659-00-9
Melting Point 242°C to 244°C (lit.)
Molecular Formula C16H16N2O3S -+ xH2O
Linear Formula C6H5NHC10H6SO3NH4 -+ xH2O
MDL Number MFCD07809526
Quantity 5 g
Molecular Weight (g/mol) 316.37 (anhydrous basis)
Percent Purity 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.