accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 206752-32-1

IUPAC Name: sodium 4-hydroxy-5-{[(6-oxocyclohexa-2,4-dien-1-ylidene)methyl]amino}-7-sulfonaphthalene-2-sulfonate
Synonyms: sodium 4-hydroxy-5-{[(6-oxocyclohexa-2,4-dien-1-ylidene)methyl]amino}-7-sulfonaphthalene-2-sulfonate
Molecular Weight (g/mol): 445.39
Molecular Formula: C17H12NNaO8S2
InChi Key: GRYBZNLNHVSHPY-UHFFFAOYSA-M
SMILES: [Na+].OC1=C2C(NC=C3C=CC=CC3=O)=CC(=CC2=CC(=C1)S([O-])(=O)=O)S(O)(=O)=O
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)