Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Azomethine-H Monosodium Salt Hydrate

Catalog No. 116355G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-635-5G 5 g
11-635-25G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-635-5G Supplier Sigma Aldrich Supplier No. 116355G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Azomethine-H Monosodium Salt Hydrate

Specifications

CAS 206752-32-1
Molecular Formula C17H12NNaO8S2 -+ xH2O
Linear Formula C17H12NNaO8S2 -+ xH2O
MDL Number MFCD00149588
Quantity 5 g
Synonym 4-Hydroxy-5-(2-hydroxybenzylideneamino)-naphthalene-2,7-disulfonic acid monosodium salt hydrate; 4-Hydroxy-5-(salicylideneamino)-2,7-naphthalenedisulfonic acid monosodium salt
Molecular Weight (g/mol) 445.4 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.