accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 206752-42-3

IUPAC Name: N-{9-[4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-9H-purin-6-yl}benzamide hydrate
Synonyms: adenosine, N-benzoyl-2'-deoxy- hydrate
Molecular Weight (g/mol): 373.37
Molecular Formula: C17H19N5O5
InChi Key: WRMYMWRRSSDKIJ-UHFFFAOYNA-N
SMILES: O.OCC1OC(CC1O)N1C=NC2=C(NC(=O)C3=CC=CC=C3)N=CN=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results