accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 20680-59-5

IUPAC Name: 3-methylbenzene-1-carboximidamide hydrochloride
Synonyms: 3-methylbenzenecarboximidamide hydrochloride
Molecular Weight (g/mol): 170.64
Molecular Formula: C8H11ClN2
InChi Key: QEAXZIMXYPAZAX-UHFFFAOYSA-N
SMILES: Cl.CC1=CC=CC(=C1)C(N)=N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results