Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methylbenzenecarboximidamide hydrochloride, 95%, Thermo Scientific™

Catalog No. SPB08395EA
Change view
Click to view available options
Quantity:
10 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
SPB08395EA 10 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SPB08395EA Supplier Thermo Scientific Chemicals Supplier No. SPB08395EA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 20680-59-5
Molecular Formula C8H11ClN2
Molecular Weight (g/mol) 170.64
MDL Number MFCD02180878
InChI Key QEAXZIMXYPAZAX-UHFFFAOYSA-N
Synonym 3-methylbenzenecarboximidamide hydrochloride, 3-methylbenzimidamide hydrochloride, 3-methyl-benzamidine hydrochloride, 3-methylbenzene-1-carboximidamide hydrochloride, benzenecarboximidamide, 3-methyl-, monohydrochloride, m-toluamidine hydrochloride, 3-methylbenzamidine, hydrochloride, 3-methylbenzenecarboxamidine, chloride, 3-methylbenzenecarboximidamide hcl, 3-carbamimidoyltoluene hydrochloride
PubChem CID 2800701
IUPAC Name 3-methylbenzenecarboximidamide;hydrochloride
SMILES CC1=CC=CC(=C1)C(=N)N.Cl

Specifications

CAS Min % 97.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Linear Formula CH3C6H4C(=NH)NH2·HCl
Quantity 10 g
Formula Weight 170.64
Percent Purity 97%
Chemical Name or Material 3-Methylbenzenecarboximidamide hydrochloride
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.