accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHOOBr

CAS RN 206860-50-6

IUPAC Name: 3-bromo-2-oxopropanoic acid
Synonyms: bromopyruvic acid
Molecular Weight (g/mol): 166.96
Molecular Formula: C3H3BrO3
InChi Key: PRRZDZJYSJLDBS-UHFFFAOYSA-N
SMILES: OC(=O)C(=O)CBr

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results