accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNCH3NNOHO(E/Z)

CAS RN 20691-84-3

IUPAC Name: 3-{2-[4-(dimethylamino)phenyl]diazen-1-yl}benzoic acid
Synonyms: 3-{2-[4-(dimethylamino)phenyl]diazen-1-yl}benzoic acid
Molecular Weight (g/mol): 269.3
Molecular Formula: C15H15N3O2
InChi Key: JAMPLPMVLCTBSB-UHFFFAOYSA-N
SMILES: CN(C)C1=CC=C(C=C1)N=NC1=CC=CC(=C1)C(O)=O
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results