Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

m-Methyl Red, TCI America™
SDP

Catalog No. M04221G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M04221G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M04221G Supplier TCI America Supplier No. M04221G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 20691-84-3
Molecular Formula C15H15N3O2
Molecular Weight (g/mol) 269.304
MDL Number MFCD00059641
InChI Key JAMPLPMVLCTBSB-UHFFFAOYSA-N
Synonym 4-Dimethylaminoazobenzene-3′C-carboxylic Acid
PubChem CID 30230
IUPAC Name 3-[[4-(dimethylamino)phenyl]diazenyl]benzoic acid
SMILES CN(C)C1=CC=C(C=C1)N=NC2=CC=CC(=C2)C(=O)O

Specifications

Quantity 1 g
Formula Weight 269.30
Physical Form Crystalline Powder
Chemical Name or Material m-Methyl Red
RTECSNumber DG8959500
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.