accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOOOOOHOAgAgAg

CAS RN 206986-90-5

IUPAC Name: trisilver(1+) 2-hydroxypropane-1,2,3-tricarboxylate
Synonyms: silver citrate
Molecular Weight (g/mol): 512.7
Molecular Formula: C6H5Ag3O7
InChi Key: QUTYHQJYVDNJJA-UHFFFAOYSA-K
SMILES: [Ag+].[Ag+].[Ag+].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O
Molecular Weight (g/mol) 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results