Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Silver citrate hydrate

Catalog No. 36125910G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
36-125-910G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 36-125-910G Supplier Sigma Aldrich Supplier No. 36125910G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Silver citrate hydrate

Specifications

CAS 206986-90-5
Melting Point 170°C
Molecular Formula C6H5Ag3O7 -+ xH2O
Linear Formula AgO2CCH2C(OH)(CO2Ag)CH2CO2Ag-+xH2O
Quantity 10 g
MDL Number MFCD00150589
Synonym Citric acid trisilver salt hydrate
Molecular Weight (g/mol) 512.7
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.