accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOSNH2

CAS RN 20699-85-8

IUPAC Name: methyl 5-amino-1-benzothiophene-2-carboxylate
Synonyms: methyl 5-amino-1-benzothiophene-2-carboxylate
Molecular Weight (g/mol): 207.25
Molecular Formula: C10H9NO2S
InChi Key: SIMBULBEQVHFPK-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC2=CC(N)=CC=C2S1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results