Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl-5-amino-1-benzothiophene-2-carboxylate, Thermo Scientific™

Catalog No. MO00936DA
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MO00936DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MO00936DA Supplier Thermo Scientific Chemicals Supplier No. MO00936DA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 20699-85-8
Molecular Formula C10H9NO2S
Molecular Weight (g/mol) 207.247
InChI Key SIMBULBEQVHFPK-UHFFFAOYSA-N
PubChem CID 2824064
IUPAC Name methyl 5-amino-1-benzothiophene-2-carboxylate
SMILES COC(=O)C1=CC2=C(S1)C=CC(=C2)N

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Synonym methyl 5-aminobenzo b thiophene-2-carboxylate, 5-aminothionaphthene-2-carboxylic acid methyl ester, methyl 5-aminobenzothiophene-2-carboxylate, benzo b thiophene-2-carboxylic acid, 5-amino-, methyl ester, maybridge3_004413, methyl 5-aminobenzo b thiophene-carboxylate, methyl5-aminobenzo b thiophene-2-carboxylate, methyl 5-azanyl-1-benzothiophene-2-carboxylate, methyl-5-amino-benzo b thiophene-2-carboxylate, methyl 5-amino-1-benzo b thiophene-2-carboxylate
Formula Weight 207.25
Percent Purity 97%
Chemical Name or Material Methyl 5-amino-1-benzothiophene-2-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.