accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 207-08-9

IUPAC Name: benzo[k]fluoranthene
Synonyms: benzo[k]fluoranthene benzo(K)fluoranthene dibenzo(b,jk)fluorene 11,12-benzo(k)fluoranthene benzo(k)fluoranthene ccris 74 2,3,1',8'-binaphthylene 8,9-benzofluoranthene 8,9-benzfluoranthene 11,12-benzofluoranthene
Molecular Weight (g/mol): 252.32
Molecular Formula: C20H12
InChi Key: HAXBIWFMXWRORI-UHFFFAOYSA-N
SMILES: C1=CC=C2C=C3C(=CC2=C1)C1=CC=CC2=CC=CC3=C12
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results