accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2070-70-4

IUPAC Name: 1,1,1,2,3,4,5,5,5-nonafluoro-4-(trifluoromethyl)pent-2-ene
Synonyms: 1,1,1,2,3,4,5,5,5-nonafluoro-4-(trifluoromethyl)pent-2-ene
Molecular Weight (g/mol): 300.05
Molecular Formula: C6F12
InChi Key: SAPOZTRFWJZUFT-UHFFFAOYSA-N
SMILES: FC(=C(F)C(F)(C(F)(F)F)C(F)(F)F)C(F)(F)F
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)