accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 207226-35-5

IUPAC Name: calcium trisodium 2-[bis({2-[bis(carboxylatomethyl)amino]ethyl})amino]acetate hydrate
Synonyms: calcium trisodium dtpa hydrate
Molecular Weight (g/mol): 515.37
Molecular Formula: C14H20CaN3Na3O11
InChi Key: AWBWYUNJDQLQGT-UHFFFAOYSA-I
SMILES: O.[Na+].[Na+].[Na+].[Ca++].[O-]C(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CCN(CC([O-])=O)CC([O-])=O
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results