Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Trimethylphosphine Solution

Catalog No. 324108100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
32-410-8100ML 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 32-410-8100ML Supplier Sigma Aldrich Supplier No. 324108100ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Trimethylphosphine is the reagent used in Mitsunobu reaction. It participates in the transformation of azides into carbamates, aziridines from azidoalcohols, iminophosphoranes and aza-Wittig reaction. Electron diffraction study of trimethylphosphine has been reported.Trimethylphosphine (PMe3) solution is the suitable reagent used in the synthesis of Fe/Te cluster type complex, Fe6Te8(PMe3)6. It may be employed as a probe to investigate the acid sites in Y-zeolite. It may be used for the synthesis of hexakis(trimethylphosphine)tris-++-methylene-diruthenium(III).

Specifications

CAS 207226-35-5
Molecular Formula C14H18CaN3Na3O10 -+ xH2O
Linear Formula C14H18CaN3Na3O10 -+ xH2O
Quantity 100 g
MDL Number MFCD00150947
Synonym PMe3; Trimethylphosphorus
Molecular Weight (g/mol) 497.35 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.