accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 207386-91-2

IUPAC Name: bis(quinolin-8-ol) sulfuric acid hydrate
Synonyms: bis(8 hydroxyquinoline) sulfuric acid hydrate
Molecular Weight (g/mol): 406.41
Molecular Formula: C18H18N2O7S
InChi Key: BNCXJZDIJIVJJO-UHFFFAOYSA-N
SMILES: O.OS(O)(=O)=O.OC1=C2N=CC=CC2=CC=C1.OC1=C2N=CC=CC2=CC=C1
Molecular Weight (g/mol) 
  • (4)
  • (2)
Percent Purity 
  • (4)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (5)
  • (4)
  • (2)
  • (4)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)