Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 8-Hydroxyquinoline hemisulfate salt hemihydrate

Catalog No. 108073100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-807-3100G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 10-807-3100G Supplier Sigma Aldrich Supplier No. 108073100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Reactant involved in synthesis of:
  • Binaphthyl-based amino acids and amino alcohols via domino coupling reactions and lactam ring opening of intermediates
  • Benzimidazoles and benzimidazolequinone derivatives via cyclocondensation
  • Axially chiral biaryls via suzuki-miyaura reactions
  • Aryl ring-fused benzimidazolequinones via radical cyclization
  • Probes for human cytochrome p450 enzymes
  • Phananthridinone derivatives via domino coupling reactions

Specifications

CAS 207386-91-2
Melting Point 176°C to 179°C (lit.)
Molecular Formula C9H7NO -+ 0.5H2O4S -+ 0.5H2O
Linear Formula C9H7NO -+ 0.5H2O4S -+ 0.5H2O
MDL Number MFCD00149610
Quantity 1 g
Synonym 8-Quinolinol hemisulfate; 8-Quinolinol sulfate (2:1) monohydrate
Molecular Weight (g/mol) 203.2
Percent Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.