accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NClNCH3H3CH3CH3CH3CI(E/Z)(E/Z)(E/Z)

CAS RN 207399-07-3

IUPAC Name: 2-(2-{2-chloro-3-[2-(3,3-dimethyl-1-propyl-2,3-dihydro-1H-indol-2-ylidene)ethylidene]cyclohex-1-en-1-yl}ethenyl)-3,3-dimethyl-1-propyl-3H-indol-1-ium iodide
Synonyms: 2-(2-{2-chloro-3-[2-(3,3-dimethyl-1-propylindol-2-ylidene)ethylidene]cyclohex-1-en-1-yl}ethenyl)-3,3-dimethyl-1-propylindol-1-ium iodide
Molecular Weight (g/mol): 667.12
Molecular Formula: C36H44ClIN2
InChi Key: IRPKBYJYVJOQHQ-UHFFFAOYSA-M
SMILES: [I-].CCCN1C2=CC=CC=C2C(C)(C)C1=CC=C1CCCC(C=CC2=[N+](CCC)C3=CC=CC=C3C2(C)C)=C1Cl
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)