Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

11-Chloro-1,1'-di-n-propyl-3,3,3',3'-tetramethyl-10,12-trimethyleneindatricarbocyanine iodide, 95%

Catalog No. AAH3215006
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3215006 5 g
AAH3215003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AAH3215006 Supplier Thermo Scientific Chemicals Supplier No. H3215006
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart
This item is not returnable. View return policy

CAS: 207399-07-3 | C36H44ClIN2 | 667.116 g/mol

11-Chloro-1,1'-di-n-propyl-3,3,3',3'-tetramethyl-10,12-trimethyleneindatricarbocyanine iodide is used as a laser and IR dye. It is also used as lithographic computer to plate laser applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
11-Chloro-1,1′-di-n-propyl-3,3,3′,3′-tetramethyl-10,12-trimethyleneindatricarbocyanine iodide is used as a laser and IR dye. It is also used as lithographic computer to plate laser applications.

Solubility
It is soluble in methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 207399-07-3
Molecular Formula C36H44ClIN2
Molecular Weight (g/mol) 667.116
MDL Number MFCD00191907
InChI Key IRPKBYJYVJOQHQ-UHFFFAOYSA-M
Synonym 2-2-2-chloro-3-1,3-dihydro-3,3-dimethyl-1-propyl-2h-indol-2-ylidene ethylidene-1-cyclohexen-1-yl ethenyl-3,3-dimethyl-1-propylindolium iodide
PubChem CID 118987185
IUPAC Name 2-[(2E)-2-[2-chloro-3-[(E)-2-(3,3-dimethyl-1-propylindol-1-ium-2-yl)ethenyl]cyclohex-2-en-1-ylidene]ethylidene]-3,3-dimethyl-1-propylindole;iodide
SMILES CCCN1C2=CC=CC=C2C(C1=CC=C3CCCC(=C3Cl)C=CC4=[N+](C5=CC=CC=C5C4(C)C)CCC)(C)C.[I-]

Specifications

Melting Point 232°C to 234°C
Quantity 5 g
Solubility Information It is soluble in methanol.
Formula Weight 667.12
Percent Purity 95%
Chemical Name or Material 11-Chloro-1,1'-di-n-propyl-3,3,3',3'-tetramethyl-10,12-trimethyleneindatricarbocyanine iodide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.