accessibility menu, dialog, popup

UserName

Viewing profile as user:
NHNOHCl

CAS RN 207403-36-9

IUPAC Name: N-[3-(azepan-1-yl)propyl]-2-cyclohexyl-2-phenylacetamide hydrochloride
Synonyms: N-[3-(azepan-1-yl)propyl]-2-cyclohexyl-2-phenylacetamide hydrochloride
Molecular Weight (g/mol): 393.01
Molecular Formula: C23H37ClN2O
InChi Key: QLMQSFIGPVLVAO-UHFFFAOYNA-N
SMILES: Cl.O=C(NCCCN1CCCCCC1)C(C1CCCCC1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results