accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 207742-76-5

IUPAC Name: pyrimidine-4,5,6-triamine; sulfuric acid
Synonyms: pyrimidine-4,5,6-triamine; sulfuric acid
Molecular Weight (g/mol): 223.21
Molecular Formula: C4H9N5O4S
InChi Key: RKJICTKHLYLPLY-UHFFFAOYSA-N
SMILES: OS(O)(=O)=O.NC1=NC=NC(N)=C1N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results