accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2078-71-9

IUPAC Name: (2-hydroxyethyl)urea
Synonyms: hydroxyethyl urea
Molecular Weight (g/mol): 104.11
Molecular Formula: C3H8N2O2
InChi Key: CLAHOZSYMRNIPY-UHFFFAOYSA-N
SMILES: NC(=O)NCCO
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results