Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Hydroxyethylurea 98.0+%, TCI America™
SDP

Catalog No. H070025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H070025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H070025G Supplier TCI America Supplier No. H070025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2078-71-9
Molecular Formula C3H8N2O2
Molecular Weight (g/mol) 104.11
MDL Number MFCD00059080
InChI Key CLAHOZSYMRNIPY-UHFFFAOYSA-N
PubChem CID 73984
IUPAC Name (2-hydroxyethyl)urea
SMILES NC(=O)NCCO

Specifications

Melting Point 93°C
Color White
Quantity 25 g
Formula Weight 104.11
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 2-Hydroxyethylurea
RTECSNumber YT4907000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.