accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 208-96-8

IUPAC Name: acenaphthylene
Synonyms: acenaphthylene cyclopenta(de)naphthalene acenaphthalene
Molecular Weight (g/mol): 152.2
Molecular Formula: C12H8
InChi Key: HXGDTGSAIMULJN-UHFFFAOYSA-N
SMILES: C1=CC2=C3C1=CC=CC3=CC=C2
Molecular Weight (g/mol) 
  • (2)
  • (4)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (4)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)