Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acenaphthylene, tech. 80%, Thermo Scientific™

Catalog No. AA3222936
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3222936 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA3222936 Supplier Thermo Scientific Chemicals Supplier No. 3222936

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 208-96-8
Molecular Formula C12H8
Molecular Weight (g/mol) 152.20
MDL Number MFCD00003806
InChI Key HXGDTGSAIMULJN-UHFFFAOYSA-N
Synonym acenaphthalene, cyclopenta de naphthalene, dsstox_cid_3845, dsstox_rid_77204, dsstox_gsid_23845, acenaphthylen, acenaphthylene, radical ion 1-, acenaphthylene
PubChem CID 9161
ChEBI CHEBI:33081
IUPAC Name acenaphthylene
SMILES C1=CC2=C3C1=CC=CC3=CC=C2

Specifications

Density 0.899
Assay Percent Range 90%
Quantity 500 g
Formula Weight 152.2
Grade Technical
Physical Form Crystalline
Chemical Name or Material Acenaphthylene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.