accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2082-66-8

IUPAC Name: {[(2-chlorophenyl)methyl]sulfanyl}carbonitrile
Synonyms: [(2-chlorophenyl)methyl]sulfanylcarbonitrile
Molecular Weight (g/mol): 183.65
Molecular Formula: C8H6ClNS
InChi Key: BEQHMRYBIXNFTP-UHFFFAOYSA-N
SMILES: ClC1=CC=CC=C1CSC#N
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results