Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chlorobenzyl Thiocyanate 98.0+%, TCI America™
SDP

Catalog No. C30035G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C30035G 5 g
C30031G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C30035G Supplier TCI America Supplier No. C30035G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2082-66-8
Molecular Formula C8H6ClNS
Molecular Weight (g/mol) 183.65
MDL Number MFCD09025671
InChI Key BEQHMRYBIXNFTP-UHFFFAOYSA-N
Synonym Thiocyanic Acid 2-Chlorobenzyl Ester
PubChem CID 286723
IUPAC Name {[(2-chlorophenyl)methyl]sulfanyl}carbonitrile
SMILES ClC1=CC=CC=C1CSC#N

Specifications

Color Yellow
Boiling Point 140°C
Quantity 5 g
Formula Weight 183.65
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Chlorobenzyl Thiocyanate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.