accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3H3COH

CAS RN 20834-59-7

IUPAC Name: 2-methyl-1-(4-methylphenyl)propan-2-ol
Synonyms: trimethylphenethyl alcohol
Molecular Weight (g/mol): 164.25
Molecular Formula: C11H16O
InChi Key: LABNAHQUILHURR-UHFFFAOYSA-N
SMILES: CC1=CC=C(CC(C)(C)O)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results