Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methyl-1-(p-tolyl)-2-propanol 98.0+%, TCI America™
SDP

Catalog No. M201925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M201925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M201925G Supplier TCI America Supplier No. M201925G

missing translation for 'validMainframeMsg'

Specifications

CAS 20834-59-7
Melting Point 42°C
Color White
Boiling Point 112°C
Molecular Formula C11H16O
Quantity 25 g
MDL Number MFCD00029997
Synonym 1,1-Dimethyl-2-(p-tolyl)ethanol, alpha,alpha,4-Trimethylphenethyl Alcohol
InChI Key LABNAHQUILHURR-UHFFFAOYSA-N
SMILES CC1=CC=C(C=C1)CC(C)(C)O
IUPAC Name 2-methyl-1-(4-methylphenyl)propan-2-ol
Molecular Weight (g/mol) 164.248
PubChem CID 88710
Formula Weight 164.25
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Methyl-1-(p-tolyl)-2-propanol
Show More Show Less
EINECSNumber (3)-1073
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.