accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 20866-46-0

OOOHOOOHNNN(S)CH3H3CCH3CH3CH3H3C

CAS RN 20866-46-0

IUPAC Name: (2S)-3-{1-[(tert-butoxy)carbonyl]-1H-imidazol-4-yl}-2-{[(tert-butoxy)carbonyl]amino}propanoic acid
Synonyms: (2S)-2-[(tert-butoxycarbonyl)amino]-3-[1-(tert-butoxycarbonyl)imidazol-4-yl]propanoic acid
Molecular Weight (g/mol): 355.39
Molecular Formula: C16H25N3O6
InChi Key: IXHPIPUIOSSAIS-NSHDSACASA-N
SMILES: CC(C)(C)OC(=O)N[C@@H](CC1=CN(C=N1)C(=O)OC(C)(C)C)C(O)=O
Grade 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)