accessibility menu, dialog, popup

UserName

Viewing profile as user:
NClF

CAS RN 20885-12-5

IUPAC Name: 2-chloro-6-fluoropyridine
Synonyms: 2-chloro-6-fluoropyridine
Molecular Weight (g/mol): 131.53
Molecular Formula: C5H3ClFN
InChi Key: LXOHKRGLGLETIJ-UHFFFAOYSA-N
SMILES: FC1=NC(Cl)=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results