Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-6-fluoropyridine 98.0+%, TCI America™
SDP

Catalog No. C31865G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C31865G 5 g
C318625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C31865G Supplier TCI America Supplier No. C31865G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 20885-12-5
Molecular Formula C5H3ClFN
Molecular Weight (g/mol) 131.534
MDL Number MFCD00077483
InChI Key LXOHKRGLGLETIJ-UHFFFAOYSA-N
PubChem CID 639437
IUPAC Name 2-chloro-6-fluoropyridine
SMILES C1=CC(=NC(=C1)Cl)F

Specifications

Melting Point 33°C
Color White
Quantity 5 g
Formula Weight 131.53
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2-Chloro-6-fluoropyridine
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.