accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3CH3OONH(R)SHOHO

CAS RN 20887-95-0

IUPAC Name: (2R)-2-{[(tert-butoxy)carbonyl]amino}-3-sulfanylpropanoic acid
Synonyms: (2R)-2-[(tert-butoxycarbonyl)amino]-3-sulfanylpropanoic acid
Molecular Weight (g/mol): 221.27
Molecular Formula: C8H15NO4S
InChi Key: ATVFTGTXIUDKIZ-YFKPBYRVSA-N
SMILES: CC(C)(C)OC(=O)N[C@@H](CS)C(O)=O
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)