CAS RN 2088828-99-1
CAS RN 2088828-99-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Promotions Available
Sigma Aldrich 3-(3-(But-3-Yn-1-Yl)-3H-Diazirin-3-Yl)Propanoic Acid
Sigma Aldrich 3-(3-(But-3-Yn-1-Yl)-3H-Diazirin-3-Yl)Propanoic Acid
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.