accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2091-46-5

IUPAC Name: triphenyl(prop-2-yn-1-yl)phosphanium bromide
Synonyms: triphenyl(prop-2-yn-1-yl)phosphanium bromide
Molecular Weight (g/mol): 381.25
Molecular Formula: C21H18BrP
InChi Key: AFZDAWIXETXKRE-UHFFFAOYSA-M
SMILES: [Br-].C#CC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)