Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triphenylpropargylphosphonium Bromide 98.0+%, TCI America™
SDP

Catalog No. P14385G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P14385G 5 g
P143825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P14385G Supplier TCI America Supplier No. P14385G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2091-46-5
Molecular Formula C21H18BrP
Molecular Weight (g/mol) 381.25
MDL Number MFCD00011811
InChI Key AFZDAWIXETXKRE-UHFFFAOYSA-M
Synonym triphenylpropargylphosphonium bromide, propargyltriphenylphosphonium bromide, prop-2-ynyltriphenylphosphonium bromide, triphenyl prop-2-yn-1-yl phosphonium bromide, propargyl triphenylphosphonium bromide, acmc-1cbag, propargyltriphenylphosphoniumbromide, triphenylpropargylphosphoniumbromide, triphenylprop-2-ynylphosphine, bromide, triphenyl-2-propynylphosphonium bromide
PubChem CID 2724214
IUPAC Name triphenyl(prop-2-yn-1-yl)phosphanium bromide
SMILES [Br-].C#CC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Color White
Quantity 5 g
Formula Weight 381.25
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Triphenylpropargylphosphonium Bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.