accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 20925-27-3

IUPAC Name: 4-amino-2-chlorobenzonitrile
Synonyms: 4-amino-2-chlorobenzonitrile
Molecular Weight (g/mol): 152.58
Molecular Formula: C7H5ClN2
InChi Key: ZFBKYGFPUCUYIF-UHFFFAOYSA-N
SMILES: NC1=CC=C(C#N)C(Cl)=C1
Percent Purity 
  • (3)
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)