Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Amino-2-chlorobenzonitrile, 99%

Catalog No. AC443880100
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC443880100 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC443880100 Supplier Thermo Scientific Chemicals Supplier No. 443880100
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 20925-27-3 | C7H5ClN2 | 152.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20925-27-3
Molecular Formula C7H5ClN2
Molecular Weight (g/mol) 152.58
MDL Number MFCD00035926
InChI Key ZFBKYGFPUCUYIF-UHFFFAOYSA-N
Synonym 3-chloro-4-cyanoaniline, 2-chloro-4-aminobenzonitrile, benzonitrile, 4-amino-2-chloro, 4-amino-2-chloro-benzonitrile, 4-amino-2-chlorobenzenecarbonitrile, pubchem4620, 4-cyano-3-chloroaniline, acmc-1cfb7, 3-chloro-4-cyanobenzenamine, ksc497i5p
PubChem CID 88728
IUPAC Name 4-amino-2-chlorobenzonitrile
SMILES NC1=CC=C(C#N)C(Cl)=C1

Specifications

Melting Point 116.0°C to 118.0°C
Quantity 10 g
Packaging Glass bottle
Assay Percent Range 98.5% min. (HPLC)
Solubility Information Solubility in water: insoluble
Formula Weight 152.58
Percent Purity 99%
Chemical Name or Material 4-Amino-2-chlorobenzonitrile
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
EINECSNumber 244-114-

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.