accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2094-99-7

CH3CH3NOCH3H2C

CAS RN 2094-99-7

IUPAC Name: 3-(prop-1-en-2-yl)-N-(propan-2-ylidene)benzamide
Synonyms: 3-(prop-1-en-2-yl)-N-(propan-2-ylidene)benzamide
Molecular Weight (g/mol): 201.27
Molecular Formula: C13H15NO
InChi Key: GSXKEHUTLZDROA-UHFFFAOYSA-N
SMILES: CC(C)=NC(=O)C1=CC=CC(=C1)C(C)=C
Boiling Point 
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)