Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3-Isopropenyl-alpha,alpha-dimethylbenzyl isocyanate

Catalog No. 361771250ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
250 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
36-177-1250ML 250 mL
36-177-11L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 36-177-1250ML Supplier Sigma Aldrich Supplier No. 361771250ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

3-Isopropenyl-α,α-dimethylbenzyl isocyanate (m-TMI) is a telechelic derivative that shows anα-unsaturation and has an isocyanate end group. It is majorly used in polymeric synthesis.m-TMI can be used as a tetrafunctional isocyanate which can further be used in the development of polymer electrolyte. It can be used as a coupling agent in the preparation of jute reinforced polypropylene.

Specifications

CAS 2094-99-7
Density 1.018 g/mL (at 25°C)
Boiling Point 268°C to 271°C
Molecular Formula C13H15NO
Refractive Index n20/D 1.53 (literature)
Linear Formula H2C=C(CH3)C6H4C(CH3)2 NCO
Quantity 250 mL
MDL Number MFCD00080490
Synonym 3-Isopropenyl-alpha,alpha-dimethylbenzyl isocyanate; m -Isoprenyl-alpha,alpha-dimethylbenzyl isocyanate
Molecular Weight (g/mol) 201.26
Percent Purity 95%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.