CAS RN 2095485-22-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ BTD S-sulfenic acid probe
Chemoproteomic reagent for targeting S-sulfenic acids on SulfenQ platform, alkyne handle for click chemistry