accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2096330-04-8

BFONH3CCH3OHHO

CAS RN 2096330-04-8

IUPAC Name: [3-(dipropylcarbamoyl)-4-fluorophenyl]boronic acid
Synonyms: 3-(dipropylcarbamoyl)-4-fluorophenylboronic acid
Molecular Weight (g/mol): 267.11
Molecular Formula: C13H19BFNO3
InChi Key: ZOWSBHXCHLRWLC-UHFFFAOYSA-N
SMILES: CCCN(CCC)C(=O)C1=C(F)C=CC(=C1)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results