accessibility menu, dialog, popup

UserName

Viewing profile as user:
BOHNNOOFClHOOH

CAS RN 2096330-14-0

IUPAC Name: {2-chloro-4-[(2-fluoro-5-nitrophenyl)carbamoyl]phenyl}boronic acid
Synonyms: 2-chloro-4-[(2-fluoro-5-nitrophenyl)carbamoyl]phenylboronic acid
Molecular Weight (g/mol): 338.48
Molecular Formula: C13H9BClFN2O5
InChi Key: BBKOIWDLMOKXFL-UHFFFAOYSA-N
SMILES: OB(O)C1=C(Cl)C=C(C=C1)C(=O)NC1=C(F)C=CC(=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results