accessibility menu, dialog, popup

UserName

Viewing profile as user:
BOHNNOOClFHOOH

CAS RN 2096330-38-8

IUPAC Name: {5-[(2-chloro-4-nitrophenyl)carbamoyl]-2-fluorophenyl}boronic acid
Synonyms: 5-[(2-chloro-4-nitrophenyl)carbamoyl]-2-fluorophenylboronic acid
Molecular Weight (g/mol): 338.48
Molecular Formula: C13H9BClFN2O5
InChi Key: ZOVDSMABJRDXRX-UHFFFAOYSA-N
SMILES: OB(O)C1=C(F)C=CC(=C1)C(=O)NC1=C(Cl)C=C(C=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results