accessibility menu, dialog, popup

UserName

Viewing profile as user:
BSOONHOOH

CAS RN 2096331-97-2

IUPAC Name: [3-(dicyclohexylsulfamoyl)phenyl]boronic acid
Synonyms: 3-(dicyclohexylsulfamoyl)phenylboronic acid
Molecular Weight (g/mol): 365.3
Molecular Formula: C18H28BNO4S
InChi Key: PECBANZNZRTDHD-UHFFFAOYSA-N
SMILES: OB(O)C1=CC(=CC=C1)S(=O)(=O)N(C1CCCCC1)C1CCCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results