accessibility menu, dialog, popup

UserName

Viewing profile as user:
BOHNCH3FHOOH

CAS RN 2096332-78-2

IUPAC Name: (2-fluoro-5-{[(3-methylphenyl)methyl]carbamoyl}phenyl)boronic acid
Synonyms: 2-fluoro-5-{[(3-methylphenyl)methyl]carbamoyl}phenylboronic acid
Molecular Weight (g/mol): 287.1
Molecular Formula: C15H15BFNO3
InChi Key: XHPBFCUFHZANHQ-UHFFFAOYSA-N
SMILES: CC1=CC(CNC(=O)C2=CC(B(O)O)=C(F)C=C2)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results