accessibility menu, dialog, popup

UserName

Viewing profile as user:
BOHNNFHOOH

CAS RN 2096333-00-3

IUPAC Name: {3-fluoro-4-[(pyridin-4-yl)carbamoyl]phenyl}boronic acid
Synonyms: 3-fluoro-4-[(pyridin-4-yl)carbamoyl]phenylboronic acid
Molecular Weight (g/mol): 260.03
Molecular Formula: C12H10BFN2O3
InChi Key: AMRBHGONZSDJLK-UHFFFAOYSA-N
SMILES: OB(O)C1=CC(F)=C(C=C1)C(=O)NC1=CC=NC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results