accessibility menu, dialog, popup

UserName

Viewing profile as user:
BFONCH3HOOH

CAS RN 2096333-24-1

IUPAC Name: {3-[benzyl(ethyl)carbamoyl]-4-fluorophenyl}boronic acid
Synonyms: 3-[benzyl(ethyl)carbamoyl]-4-fluorophenylboronic acid
Molecular Weight (g/mol): 301.12
Molecular Formula: C16H17BFNO3
InChi Key: XJQIDURCDMESBR-UHFFFAOYSA-N
SMILES: CCN(CC1=CC=CC=C1)C(=O)C1=C(F)C=CC(=C1)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results